Condensers, double surface, Davies, with Quickfit® ground joints
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
C5/12/SC C5/13/SC C5/22/SC C5/23/SC C5/24/SC C5/11/SC
201-8287EA 148 EUR InStock
201-8287 201-8288 201-8289 201-8290 201-8291 201-8286
Condensers, double surface, Davies, with Quickfit® ground joints
Kühler
PYREX®, borosilicate glass.
  • Detachable plastic screw thread connector
  • High chemical and temperature resistance
Double surface, highly efficient condensers with a large cooling surface, inner cooling surface provides “baffle” effect whilst outer prevents “creep” with low boiling point liquids.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge