Condensers, coil type, with Quickfit® ground joints
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
CX3/05/SC CX3/06/SC
201-8300EA 133 EUR InStock
201-8300 201-8301
Condensers, coil type, with Quickfit® ground joints
Kühler
PYREX®, borosilicate glass.
  • Highly efficient condensers with a large cooling surface
  • Very good chemical and temperature resistance
Coil type, with ground cone. Ideal for use in the upright reflux mode where counter-current cooling is possible. Detachable plastic connectors for the attachment of rubber hose.
ISO 1773, BS 6352
Zertifizierungen: ISO 1773, BS 6352
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge