Multiple adapters, Quickfit®
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MA1/2 MA1/3 MA1/11 MA1/33
201-8572EA 78 EUR InStock
201-8572 201-8573 201-8571 201-8574
Multiple adapters, Quickfit®
Destillationsbauteile Mehrhalsaufsätze
PYREX®, borosilicate glass, straight.
  • Both sockets are the same size
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters with two parallel necks/sockets and a cone.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge