Hollow-blown glass stoppers, B-length, Quickfit®
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SB10 SB19 SB24 SB45 SB14 SB34 SB29 SB12
201-0528EA 46.9 EUR InStock
201-0528 201-8674 201-8675 201-0531 201-8672 201-2376 201-0530 BIBBSB12
Hollow-blown glass stoppers, B-length, Quickfit®
Stopfen Glasstopfen
These glass stoppers are ideally suited for use with Quickfit® jointed glassware and other glassware having standard ground glass sockets.
  • Chemical resistance
  • High temperature resistance
  • Hollow-blown construction to reduce weight
  • Hexagonal head prevents the stopper from rolling and provides a good grip
All Quickfit® stoppers conform to the standard tapers as outlined in ISO 383.
ISO 383
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge