Sorbent tubes for gas and vapor sampling, ORBO™
Lieferant: Sigma-Aldrich
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
20267-U 20257-U 20265 20349 20351 20355 20358 20360 20362 20832-U 20030-U 20044 20051 20053 20054 20063 20081-U 20211 20254-U 20255-U 20256-U 54020-U 23446-U 54081-U
20267-UEA 148 EUR InStock
20267-U 20257-U SUPL20265 20349. SUPL20351 20355. 20358. SUPL20360 20362. 20832-U 20030-U SUPL20044 SUPL20051 SUPL20053 SUPL20054 SUPL20063 20081-U SUPL20211 20254-U 20255-U 20256-U 54020-U 23446-U 54081-U
Sorbent tubes for gas and vapor sampling, ORBO™
Gasrohre
ORBO™ sorbent tubes contain two beds of the same selective adsorbent seperated by glass wool or foam, for gas and vapor sampling.
  • Wide range of high quality adsorbents
  • Consistent adsorbent mesh sizes
  • Coated and specially-treated materials
  • Reproducible recoveries and low standard deviation
  • Consistent bed weights from tube to tube to ensure capacity
The dual-layer or two-bed construction of the tube allows for any sample breakthough to be captured in the smaller back-up bed.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge