Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IgG Donkey Polyclonal Antibody (CF™ 350)

Lieferant: Biotium

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

20361-50UL
20361-50ULEA 127 EUR InStock
20361-50UL
Anti-IgG Donkey Polyclonal Antibody (CF™ 350)
Antikörper
CF™350 is a blue fluorescent dye with excitation/emission maxima at 347/448nm. CF™350 is optimized for UV excitation, and is spectrally similar to AlexaFluor®350, AMCA, and DyLight®350. CF™350 is more water soluble, more photostable, and 50% brighter when conjugated to antibodies than the traditional blue dye AMCA. CF™350 antibody conjugates are comparable in brightness to AlexaFluor®350 dye conjugates.

CF™350 is a blue fluorescent dye with excitation/emission maxima at347/448 nm. CF™350 is optimized for UV excitation, and is spectrally similar to AlexaFluor®350, AMCA, and DyLight™350. CF™350 is more water soluble, more photostable, and 50% brighter when conjugated to antibodies than the traditional blue dye AMCA. CF™350 antibody conjugates are comparable in brightness to AlexaFluor®350 dye conjugates.

CF™ dyes are covered by pending U.S. and international patents. CF™ is a trademark of Biotium, Inc. Alexa Fluor® is a registered trademark of Molecular Probes, Inc. DyLight® is a registered trademark of Pierce Biotechnology.

Bestellinformation: Antibodies are supplied at 2mg/mL in PBS pH 7.4 containing 50% glycerol, 2mg/mL BSA (IgG- and protease-free) and 0.05% sodium azide. Highly cross-adsorbed antibodies are adsorbed against whole serum protein and have minimal cross-reactivity for multiple labeling experiments as indicated. Bv: bovine; Ch: chicken; Gt: goat; GP: Guinea pig; Hs: horse; Hu: human; Ms: mouse; Rb: rabbit; Sh: sheep; SHm: Syrian hamster; Sw: swine; Rt: rat.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF350
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge