Glass wool, Supelco®
Lieferant: Sigma-Aldrich
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
20383 20384 20409 20410 20411 21688-U
20383.EA 73 EUR InStock
20383. 20384. 20409. 20410. 20411. 21688-U
Glass wool, Supelco®
Glas- und Baumwolle
Wool plugs are often used in GC inlet liners to improve sample vaporization, and/or to keep non-volatile material from entering the column. They can also be used in packed GC columns, solvent desorption tubes, thermal desorption tubes, and purge traps to retain packed beds.
Silanised: For general purpose use
Pesticide Grade (Silanised): For applications that involve active analytes, such as organochlorine and/or organophosphorous pesticides.
Phosphoric acid treated: For acidic compounds such as barbiturates, free fatty acids, etc.
Non-treated: For special application where the user deactivates based on their intended analytes
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge