Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IgG Fc gamma Goat Polyclonal Antibody (CF™ 640R)

Lieferant: Biotium

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

20410-500UG
20410-500UGEA 285 EUR InStock
20410-500UG
Anti-IgG Fc gamma Goat Polyclonal Antibody (CF™ 640R)
Antikörper
This product is prepared by labeling F(ab’)2 fragment goat anti-mouse IgG (H+L), Fcgamma fragment specific, with CF™640R. The antibody has been adsorbed against human, bovine and horse serum proteins to minimize cross-reactivity. Far-red fluorescent CF™640R (Ex/Em 642/662 nm) is much brighter than Cy®5 and at least as bright as Alexa Fluor® 647, and has exceptional photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Secondary
Antigen: IgG Fc gamma
Clonality: Polyclonal
Clone:
Conjugation: CF640R
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge