May-Grünwalds Eosin-Methylenblau zur Zellfärbung, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
Achtung
M-Clarity™ Qualitätsstufe = MQ200
205435-25G
205435-25GEA 103 EUR InStock
205435-25G
May-Grünwalds Eosin-Methylenblau zur Zellfärbung, Sigma-Aldrich®
May-Grünwalds Eosin-Methylenblau
May-Grünwald stain is a Romanowsky neutraldye stain based on the combination ofmethylene blue azures as the basic dyecomponent and eosin as the acid dyecomponent.It is intended for use in staining blood films orbone marrow films. It is used routinely for diagnostic purposes in hematology.
  • May-Grünwald′s stain is used for the differential staining of cellular elements of blood
Order Now
Spezifikation Testergebnisse
Appearance (Colour) Green to Black
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
UV Absorbance Ratio
A650/A523
≥ 1.45
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge