20651.03
20651.06
20651.03EA
64.1
EUR
InStock
20651.03
20651.06
Octylphosphonsäure ≥98%
Octylphosphonic acid
Purity: 98%
CAS No.: 4724-48-5
Molecular Formula: C8H19O3P
Formula Weight: 194.21
Physical Form: Crystalline
Melting Point: 100-102°
Beilstein Reference: 1764242
MDL No.: MFCD00015841