Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

L(+)-Glutamin 98,5-101,5 Dry_basis USP, multi-compendial, J.T.Baker®

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


M.O.C. Category R

2078-09EA 0 EUR InStock
2078-09
L(+)-Glutamin 98,5-101,5 Dry_basis USP, multi-compendial, J.T.Baker®
L(+)-Glutamin


Formel: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Siedepunkt: 355
Melting Pt: 185…186 °C
Dichte: 1.5326 g/cm³ (130 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00008044
CAS-Nummer: 56-85-9
EINECS: 200-292-1
Merck Index: 13,04484
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&vid=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Meets U.S.P Requirements
Meets J.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₅H₁₀N₂O₃) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotation [Å]²º^D (+) 6.3 - 7.3 Degree
USP - Loss on Drying at 105°C ≤ 0.3 %
USP - Residue after Ignition ≤ 0.3 %
USP - Chloride (Cl) ≤ 0.05 %
USP - Sulfate (SO₄) ≤ 0.03 %
USP - Iron (Fe) ≤ 0.003 %
USP - Related Compounds 0.5% max
JP - Assay (C₅H₁₀N₂O₃) (dried basis) 99.0 - 101.0 %
JP - Identification Passes Test
JP - Specific Rotation [Å]²º^D (+) 6.3 - 7.3 Degree
JP - Loss on Drying at 105°C ≤ 0.30 %
JP - Residue after Ignition ≤ 0.1 %
JP - Chloride (Cl) ≤ 0.021 %
JP - Sulfate (SO₄) ≤ 0.028 %
JP - Iron (Fe) ≤ 10 ppm
JP - Heavy Metals (as Pb) ≤ 10 ppm
JP - Related substances Passes Test
JP - pH 4.5 - 6.0
JP - Clarity and Color of Solution Passes Test
JP - Ammonium ≤ 0.1 %
Solubility of 3% Solution at 30°C Passes Test
Solubility of 3.5% Solution at 30°C Passes Test
Identification (by IR) Passes Test
Endotoxin Concentration (EU/mg) = 0.06
Endotoxin Concentration, IU/mg Passes Test
No Class 1,2,3 or other solvents are used or produced in the Passes Test
manufacturing or purification of the product. = 0.3 %
FCC - Specific Rotation [Å]²⁰ D (+) 6.3 - 7.3 Degree
Elemental Impurities (USP (232), EP 5.20) - Information on elemental Passes Test
impurities for this product is available on the associated Product Passes Test
Regulatory Data Sheet and elemental impurity profile report. Passes Test

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge