20837.04
20837.09
20837.18
20837.04EA
31.3
EUR
InStock
20837.04
20837.09
20837.18
Natriumtrifluormethansulfonat
Natriumtrifluormethansulfonat
CAS No.: 2926-30-9
Molecular Formula: CF3NaO3S
Formula Weight: 172.06
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 253-255°
Beilstein Reference: 3728797
MDL No.: MFCD00061607