Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Human pooled serum, French origin

Lieferant: SEQENS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

21000P-500ML 21000P-100ML 21000P-1L
21000P-500MLEA 1970 EUR InStock
21000P-500ML 21000P-100ML 21000P-1L
Human pooled serum, French origin
Seren
This human pooled serum is obtained from off-the-clot serum units collected in France. Units are pooled and 0,2 μm filtered prior to being bottled. Possibility of gender and blood group selection.

  • Dry extract: 70 to 90 g/L
  • Total proteins: 60 to 80 g/L
  • Filtered through a 0,2 µm membrane
  • Shelf life: 3 years

Donations used for the manufacture of this product have been collected in EU licensed donation facilities, in accordance with ethical requirements of the French Code de la santé publique:
Donor consent: donations proceed from volunteer donors, who agreed their donation may be used in any way the collection centre deems appropriate
Gratuitousness: donations have not been paid
Anonymity: each donation is anonymous and is associated with a donation number

Each unit has also been tested and found negative or non-reactive for:
Anti-HIV 1+2 and anti-HCV antibodies
HBs antigen
HIV 1 and HCV RNA
Syphilitic serology

Complete donor traceability on file.

Achtung! Bitte beachten: Only for research or manufacture of in vitro diagnostic reagents. Not for therapeutic use. This product of human origin should be considered as potentially infectious and handled accordingly.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge