Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

D(+)-Galactose ≥98% (durch HPLC), Biotechreagenz, J.T.Baker®

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


2102-07EA 0 EUR InStock
2102-07
D(+)-Galactose ≥98% (durch HPLC), Biotechreagenz, J.T.Baker®
D(+)-Galactose


Formel: C₆H₁₂O₆
MW: 180.16 g/mol
Siedepunkt: 527
MDL Number: MFCD00151230
CAS-Nummer: 59-23-4
Merck Index: 13,04356
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=508b1d21a6801ddfb1a5f13a64cb06e6f95e8922&vid=508b1d21a6801ddfb1a5f13a64cb06e6f95e8922&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
GMP Manufactured Product
Arsenic (As) ≤0,500 ppm
Assay (by HPLC) ≥98%
Glucose ≤2,0%
Heavy Metals (as Cu) ≤10 ppm
Iron (Fe) ≤5 ppm
Endotoxin Concentration (EU/g) ≤10
Residual Methanol ≤3000 ppm
Residual Ethanol ≤5000 ppm
EP - Appearance of Solution Passes Test
EP - Acidity or Alkalinity Passes Test
EP - Identification A Passes Test
EP - Specific Optical Rotation [Å]²º^D (+) 78,0 - 81,5 Degree
EP - Barium (Ba) Passes Test
EP - Lead (Pb) ≤0,5 ppm
EP - Water (H₂O) ≤1,0%
EP - Ash (sulphated) ≤0,1%
EP - Total Viable Count (per g) ≤100
Only Class 2 (Methanol) and Class 3 (Ethanol) residual solvents are present. Both are tested, quantified, and reported.

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge