Racks for micro sample test tubes, PP, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F188410000
211-7420EA 105 EUR InStock
211-7420
Racks for micro sample test tubes, PP, SP Bel-Art
Gestelle Reagenzglasgestelle
PP, white.
  • Very good chemical resistance
  • Very good temperature resistance
These test tube racks stack to conserve space, even when filled with tubes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge