Calcium ionophore II for ion selective electrodes, Selectophore™, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
21193-250MG-F 21193-50MG-F
21193-250MG-FEA 831 EUR InStock
21193-250MG-F 21193-50MG-F
Calcium ionophore II for ion selective electrodes, Selectophore™, Supelco®
Elektrolyte Elektrodenfülllösungen
Lipophilic ionophore for Ca²⁺ selective electrodes with detection limit in the sub-nanomolar range.
Extremely efficient carrier of Ca²⁺ -ions; induces a selectivity in membranes for Ca²⁺ over Mg²⁺ by a factor of about 10⁴. Calcium ionophore II (ETH 129) was used in preparing calcium selective minielectrodes which are used in determination of dissociation constant (Kd).
Verpackung: Packed in bottomless glass bottle. Contents are inside inserted fused cone.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79dce6fa0178eb2f82e1641265ee048bc95a06bb&vid=79dce6fa0178eb2f82e1641265ee048bc95a06bb&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge