Test tubes, with rim, medium wall
Lieferant: MURRAY S
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
G022/31 G022/33 G022/34 G022/35 G022/38 G022/40 G022/32
212-0560EA 41.1 EUR InStock
212-0560 212-0456 212-0561 212-0562 212-0563 212-0564 212-0455
Test tubes, with rim, medium wall
Röhrchen Reagenzgläser- und Probenröhrchen
Neutral glass, SAMCO '50'
  • Medium walled
  • Withstand moderate temperature change
Zertifizierungen: Manufactured to the dimensions, tolerances and quality requirements of ISO 4142:2002, to meet all applications for re-usable glass tubes.
Achtung! Bitte beachten: Tubes should not be taken from cold and placed directly into the hottest part of the flame without pre-heating.
Order Now
PRODUKT
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge