Centrifuge tubes, with screw cap
Lieferant: GLASWARENFABRIK KARL HECHT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
40970010
212-0733EA 96.7 EUR InStock
212-0733
Centrifuge tubes, with screw cap
Röhrchen Zentrifugenröhrchen
AR®-Glass
  • Ungraduated
  • Good chemical resistance
Conical bottom, with thread GL 14 and screw cap of polypropylene.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge