VWR®, Test Tubes with Rim
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
212-0753EA 29.3 EUR InStock
212-0753 212-0754 212-0755 212-0756 212-0757 212-0758 212-0759 212-0760 212-0761 212-0762 212-0763 212-0764 212-0765
VWR®, Test Tubes with Rim
Röhrchen Reagenzgläser- und Probenröhrchen
Test tubes made from borosilicate 5.1 glass, neutral glass.
  • Thin walled
  • Excellent chemical resistance
  • Relatively resistant to rapid temperature changes and local heating
The economical alternative for a wide range of uses.
Zertifizierungen: DIN 12395.
Order Now
PRODUKT
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge