Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

MicroCNX™ connectors

Lieferant: CPC COLDER

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

CNX17001HT CNX17002HT CNX17003HT
212-0790EA 791 EUR InStock
212-0790 212-0791 212-0792
MicroCNX™ connectors
Fittings und Verbinder
Aseptic micro-connectors that provide a simple, efficient method of connecting tubing for small format biomanufacturing assemblies. MicroCNX™ connectors are the modern alternative to the cumbersome, industrial process of tube welding.

  • Pinch – click and pull: Intuitive three-step connection process reduces risk of operator error
  • Easy to use: Lowers risk of operator error and related performance, reliability and safety concerns
  • Genderless: Eases single-use systems specifications with one part number for both halves
  • CPC click: Audible confirmation of assembly with no additional hardware required

The MicroCNX™ line is engineered specifically for the challenging conditions of biologic media transfer in bioprocessing, cell therapy and gene therapy applications.

Autoclave: One cycle up to 130 °C (266 °F) for 60 minutes
Sterilisation: Up to 50 kGy gamma irradiation

Main components: Polycarbonate (white); USP Class VI, ADCF
Seals: Silicone (clear), platinum-cured; USP Class VI, ADCF
Protective cover: Polypropylene (teal); USP Class VI, ADCF
Membrane: Hydrophobic polyethersulphone; USP Class VI

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge