VWR® Culture Tubes With Screw Thread
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
212-0850EA 81.5 EUR InStock
212-0850
VWR® Culture Tubes With Screw Thread
Röhrchen Kulturröhrchen
Reusable culture tubes with screw thread made from clear borosilicate glass.
  • High resistance against chemicals
  • High thermal resistance
  • With white marking field
Bestellinformation: Screw caps have to be ordered separately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge