Test tubes, flat bottom, with screw cap
Lieferant: SEMADENI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2281
212-3120EA 68.6 EUR InStock
212-3120
Test tubes, flat bottom, with screw cap
Röhrchen Reagenzgläser- und Probenröhrchen
PS, clear, with screw cap.
  • Flat bottom
  • Good thermal resistance
  • Good chemical resistance
Universal tubes for a wide range of applications.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge