Test tubes without rim, heavy wall
Lieferant: MURRAY S
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
G033/01 G033/02 G033/03 G033/05 G033/08 G033/10 G033/06
212-7039EA 78.5 EUR InStock
212-7039 212-7040 212-7041 212-7042 212-7043 212-7044 212-0463
Test tubes without rim, heavy wall
Röhrchen Reagenzgläser- und Probenröhrchen
Borosilicate glass, grade 3.3, SAMCO '33'. Thick walled.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Suitable for all usual laboratory applications. They will withstand temperature change up to and including boiling of samples.
Zertifizierungen: ISO 4142:2002
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge