Test Tubes with Rim
Lieferant: MURRAY S
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
G001/82 G001/71 G001/72 G001/73 G001/75 G001/76 G001/80 G001/81 G001/83 G001/84 G001/79
212-0556EA 84.1 EUR InStock
212-0556 212-7076 212-7077 212-7078 212-7079 212-7080 212-7081 212-7082 212-7083 212-7084 212-0555
Test Tubes with Rim
Röhrchen Reagenzgläser- und Probenröhrchen
Soda-lime glass test tubes are suitable for general mixing and simple laboratory work, and will withstand moderate warming (water bath).
Achtung! Bitte beachten: They should not be placed directly into naked flames.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge