Bottle and vial racks, No-Wire™, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F185140025 F185140013 F185140016 F185140020 F185140030
212-8513EA 63 EUR InStock
212-8513 212-8510 212-8511 212-8512 212-8514
Bottle and vial racks, No-Wire™, SP Bel-Art
Gestelle Flaschengestelle
Racks made of PP, white.
  • Five grid configuration
  • Holds small vials and bottles
  • Autoclavable
Small enough to hold in one hand and enough choices to accommodate most vials and small bottles.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge