Microtube racks
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M-450
212-8940EA 30.5 EUR InStock
212-8940
Microtube racks
Gestelle Reagenzglasgestelle
Racks, made of PP, designed for long term storage of samples at temperatures down to −90 °C.
  • With lift off telescopic lid
  • Lids with moulded alphanumeric identification system
  • Good chemical resistance
The four frosted side panels and a frosted rim on the edges of the lid provide additional labelling options.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge