Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

PCR manual workstation

Lieferant: DELTALAB

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

M-540 M-542 M-541
212-9154EA 137 EUR InStock
212-9154 212-9156 212-9216
PCR manual workstation
Gestelle Reagenzglasgestelle
This storage/working rack, with lid, is designed to work with different PCR volumes at the same time. It can accommodate up to 32 individual 0,2 ml PCR tubes, 24× 1,5/2,0 ml and 16× 0,5 ml microtubes.

  • Tube wells are alphanumerically matrixed with raised bold type for accurate sample identification
  • Rack has a transparent lid for easy viewing of samples
  • Frosted writing panels on four sides allow labelling or writing
  • Supplied with a removable rack accommodating 0,2 ml PCR tubes

Main rack W×D×H: 215×118×50 mm
Removable rack W×D×H: 125×85×15 mm

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge