Bottles, narrow neck, round, PP, with screw caps, Azlon®
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BNP0125PN BNP0250P BNP0500P BNP1000P BNP2000P BNP0150PN
215-0529EA 23.6 EUR InStock
215-0529 215-0530 215-0531 215-0532 215-0533 215-3109
Bottles, narrow neck, round, PP, with screw caps, Azlon®
Flaschen Kunststoffflaschen
PP, durchscheinend mit PP-Schraubverschluss.
  • Very good chemical resistance
  • Autoclavable
  • Robust, leakproof
  • Heavy-duty and durable
Ideal for liquid storage, media preparation and sampling applications.
Zertifizierungen: Suitable for food contact applications in accordance with EU and U.S. FDA regulations.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge