Wash bottles, slope shoulder, Azlon®
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WGF531P WGF537P WGF536P WGF530P WGF535P WGF541P WGF539P WGF600P WGF533P WGF534P WGF540P WGF532P WGF601P WGF538P
215-3149EA 34.8 EUR InStock
215-3149 215-0582 215-0579 215-0580 215-0581 215-0583 215-0401 215-3171 215-3372 215-3374 215-3376 AZLOWGF532P 215-3377 215-4052
Wash bottles, slope shoulder, Azlon®
Flaschen Spritzflaschen
LDPE translucent, colour coded PP closure with jet-tip aperture.
  • Indelibly printed with a choice of popular laboratory solvents, appropriate hazard symbols and CAS numbers
  • Slim design offers comfortable handling and enhanced ease of use
  • Jet tip aperture for fine control
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge