Verpackungsflaschen, Enghals, rund, HDPE, mit Gewinde, Nalgene®
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
362089-0001 362089-0004 362089-0002 362089-0008 362089-0016 362089-0032
215-1234EA 582 EUR InStock
215-1234 215-1235 215-1587 215-1588 215-1589 215-1590
Verpackungsflaschen, Enghals, rund, HDPE, mit Gewinde, Nalgene®
Flaschen Kunststoffflaschen
HDPE, translucent.
  • Good chemical resistance
  • Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI
  • Supplied without closures; leakproof sealing with closures 215-1597/215-1252/215-1598/215-1611/215-1236/215-1247/215-1613
Economical and robust bottles for storing and transporting liquids.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge