Laborflasche, mit Gewinde, ohne Schraubverschluss, DURAN®
Lieferant: DWK Life Sciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
299016006
215-3262EA 631 EUR InStock
215-3262
Laborflasche, mit Gewinde, ohne Schraubverschluss, DURAN®
Flaschen Metallflaschen
Manufactured from corrosion resistant AISI Type 316 L (1.4404) stainless steel, and hygienically constructed with all welded seams.
  • Hazardous materials laboratory container for storage of liquids, intermediates, or solid product
  • Storage of precious materials, such as high purity fine chemicals, pharmaceutical or cosmetic products
  • Storage of light sensitive materials
The unbreakable steel bottle is ideal for storage applications where glass is not applicable, due to the risk of breakage or chemical incompatibility. The bottle has a smooth inner surface finish that corresponds to IIIc (DIN 17441) with rounded inner edges for ease of cleaning. Polished and brushed durable exterior finish. Bottle has a GL 45 thread with a built-in pouring lip and a rim for tapmer-evident caps. Supplied without a cap, stainless steel caps are available. It is fully compatible with all the DURAN Group GL 45 caps.
Mit aufgedrucktem Retrace Code (Chargenkennung).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge