Wash bottles, integral, wide neck, Azlon®
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WGW800P
215-4162EA 41.9 EUR InStock
215-4162
Wash bottles, integral, wide neck, Azlon®
Flaschen Spritzflaschen
Wide neck wash bottles with red, yellow, blue, green and orange PP caps.
  • Fitted with colour coded one-piece PP closure that maintains it's shape in use
  • Designed with integral spout to prevent contamination
  • Good chemical resistance
The round iIntegral wash bottle is moulded with an integral spout so you can say goodbye to the contamination caused by the inner tube touches the benchtop during re-filling.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge