Instrumentenschalen mit Deckel
Lieferant: BOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
8400 8404 8406 8407
216-0016EA 72 EUR InStock
216-0016 216-0017 216-0018 216-0019
Instrumentenschalen mit Deckel
Schalen/Schüsseln/Töpfe Instrumentenschalen
Stainless steel, corrosion-resistant, 18/10.
  • Special corner shape for fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance, very good thermal resistance
With a cover that enables the instrument dishes to be stacked.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge