VWR® Kühlakkus
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
216-1168EA 214 EUR InStock
216-1168
VWR® Kühlakkus
Kühlakkus
Long lasting, high performance temperature protection.
  • Hard plastic bottle pack is filled with water
  • Can be refrigerated or frozen to keep products cold or frozen
  • Suitable for pharmaceuticals, clinical trials, diagnostic kits, chemicals, and biologicals
Tough, resuable bottle packs are designed to safeguard temperature-sensitive products during storage and transport. They can be chilled in a freezer, refrigerator, or low temperature incubator, and are effective when used with insulated shipping containers. Phase changes at 0 °C and returns to liquid state after thawing.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge