Buckets, with leaktight lid, EUROTUBO®
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
222802 222803 222804 222805
216-1651EA 9.5 EUR InStock
216-1651 216-1652 216-1653 216-1654
Buckets, with leaktight lid, EUROTUBO®
Eimer
These disposable buckets are made of polypropylene with tight fitting and leakproof lids.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
All buckets are supplied with a sturdy and convenient white plastic handle for easier carrying.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge