VWR®, Filling and Venting Cap for VWR® Carboys
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
216-1842EA 73.5 EUR InStock
216-1842
VWR®, Filling and Venting Cap for VWR® Carboys
Verschlüsse Safety Caps und Safety Waste Caps
The filling and venting cap for VWR® carboys is an autoclavable closure designed for the aseptic transfer of media, biological reagents, pure water and chemicals
Each cap is equipped with integrated 6,35 mm ports.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge