Tanks, zylindrisch, mit Deckel, Nalgene®
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
11200-0005 11200-0007 11200-0010 11200-0015 11200-0030 11200-0055 11200-0100
216-1900EA 331 EUR InStock
216-1900 216-1901 216-1902 216-1903 216-1904 216-1905 216-1906
Tanks, zylindrisch, mit Deckel, Nalgene®
Tanks Tanks, zylindrisch
These cylindrical polypropylene tanks with cover can resist stress cracking and increase tolerance to many organic chemicals.
  • An economic alternative to stainless-steel tanks
Bestellinformation: Factory-installed spigots and tank liners are sold separately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge