Emballages pour le transport de produits infectieux
Lieferant: CORTEX AIREMBAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
56CRYOPACK12L 56CRYOPACK4L 56CRYOPACK6L 371BIOT.25ST
216-2037EA 933 EUR InStock
216-2037 216-2039 216-2040 CORT371BIOT.25ST
Emballages pour le transport de produits infectieux
Versandtaschen und Transportbehälter
Micro-fluted cardboard treated against moisture (Biotainer); outer double fluted carton (Cryopack®).
  • Cardboard wedge
  • Sheet of absorbent cloth
  • Watertight secondary PP container
These packages are designed for the transport of infected or contaminated consumables. They are suitable for applications in room temperature (Biotainer®). They can also be used in controlled temperature of 1 to 8 °C or in freezing temperatures with dry ice (Cryopack®).
Approved by the UN. Comply with international regulations in force IATA class 6.2 IMDG and ADR RIG.
Zertifizierungen: Approved by the UN. Comply with international regulations in force IATA class 6.2 IMDG and ADR RIG.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge