Attached Lid Containers
Lieferant: MULTIROIR
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BA12145 BA12146 BA12147 BA12148
216-2248EA 48.4 EUR InStock
216-2248 216-2249 216-2251 216-2252
Attached Lid Containers
Boxen Transportboxen
These containers, with robust attached lids, are designed to stack when full and nest when empty. The recesses of the hinged lid ensure high stability.
  • Made of of heavy-duty polypropylene, rigid, robust and practical
  • Containers nest for maximum space usage with lids open
  • Blue
  • When empty, saves up to 75% valuable storage space
  • Easy to clean interiors
The attached lid containers can be secured with plastic seals and transported with trolleys.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge