Single wafer shippers, for 5" (125 mm) wafer handling, H 22 series
Lieferant: ENTEGRIS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H22-501-62C02 H22-502-0615
216-2353EA 0 EUR InStock
216-2353 216-2354
Single wafer shippers, for 5" (125 mm) wafer handling, H 22 series
Wafer-Handling-Zubehör Wafer-Versandbehälter
Conical pocket holds one wafer face down contacting only the edge of the wafer.
  • Spring retains wafer and minimises particle generation
  • Stackable for efficient use of storage space
  • Loose or lock stacking, either tray on tray or cover on tray
  • Convenient screw on covers
  • Large area on cover provides space for labelling
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge