Dishes, flat bottom, with spout
Lieferant: AGENCINOX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
312A 316A 320A 325A 306 308 310 312 304
216-2715EA 56.3 EUR InStock
216-2715 216-2716 216-2717 216-2718 216-2711 216-2712 216-2713 216-2714 AGEN304
Dishes, flat bottom, with spout
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Stainless steel 18/10.
  • Without handles
  • Good chemical resistance
  • Excellent thermal resistance
Round multipurpose dishes for a wide range of uses.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge