Ointment jars, with thread
Lieferant: RIXIUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1-0900-005051I 1-0900-0030-46 1-0900-0100-62 1-0901-0100-52
216-2983EA 139 EUR InStock
216-2983 216-2972 216-2981 216-2979
Ointment jars, with thread
Dosen Dosen, Universal
Made from clear or amber soda-lime glass.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • With round thread
Ideal for sampling, storage and transportation of powders and pastes. Amber jars to protect light-sensitive materials.
Bestellinformation: Screw caps have to be ordered separately.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge