Crucibles, Vitreosil®
Lieferant: TECHNICAL QUARTZ SOLUTIONS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
73081 73082 73083
216-4496EA 87.9 EUR InStock
216-4496 216-4497 216-4498
Crucibles, Vitreosil®
Tiegel
These crucibles are made from fused quartz, translucent, glazed.
  • Max. temperature: 1050 °C
Lieferumfang: Supplied without lid.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge