Hähne für Ballons
Lieferant: hünersdorff GmbH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
520100 520700 520705 520105
216-5291EA 21.4 EUR InStock
216-5291 216-5292 HUEN520705 HUEN520105
Hähne für Ballons
Hähne
Made of fracture-proof and food-safe polyethylene with sleeve connection for a hose.
  • Very resistant to acids, alkalis and other chemicals
  • allow for extra-precise dosing
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge