Autoclavable Bags
Lieferant: SEMADENI
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2054
216-7350EA 91 EUR InStock
216-7350
Autoclavable Bags
Beutel Entsorgungsbeutel
This yellow bag is made from thick sheets of 0,05 mm. Autoclave with open sachets and to be closed again. The bags are made and stained in a non-pulling material and their combustion is carried out without risk.
  • Instructions printed in black
  • Maximum autoclave temperature: 121 °C
  • Enable easy, safe and harmless disposal of biologically or other contaminated wastes
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge