Stoppers, with drip tip, MBL®
Lieferant: SciLabware
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TRF691 TRF692 TRF693 TRF695 TRF696 TRF697 TRF690 TRF698
217-0745EA 49.2 EUR InStock
217-0745 BIBBTRF692 217-0746 217-0747 217-0748 217-0749 BIBBTRF690 BIBBTRF698
Stoppers, with drip tip, MBL®
Stopfen Glasstopfen
Ideally suited for use with volumetric flasks etc. Lower closed end of stopper features a 'drip-tip' to ensure adhered liquid is returned to the flask. Flat head allows stopper to be stood on end, thus preventing contamination of the ground surface.
  • Hexagonal head prevents the stopper from rolling and provides a good grip
  • Hollow-blown to reduce weight
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge