Stoppers for Conical Ground Joints
Lieferant: Lenz Laborglas GmbH & CO.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
01400512 01400529 01400514 01400519
217-9096EA 9.7 EUR InStock
217-9096 217-9099 217-9097 217-9098
Stoppers for Conical Ground Joints
Stopfen Glasstopfen
DURAN®, borosilicate glass stopper, clear, solid or semi hollow.
  • Excellent chemical resistance
  • High thermal resistance and minimal heat expansion results in high resistance to temperature changes
  • Octagonal top allows easy release and removal of the stopper
Zertifizierungen: DIN 12252
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge