CENTA™ β-Lactamase Substrate 98 HPLC_ASSAY_METHOD, powder_yellow, Millipore®
Lieferant: Calbiochem (Part of Merck)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
219475-25
219475-25EA 358 EUR InStock
219475-25
CENTA™ β-Lactamase Substrate 98 HPLC_ASSAY_METHOD, powder_yellow, Millipore®
CENTA™ β-Lactamase Substrate
CENTA™ β-lactamase substrate, is a chromogenic β-lactamase substrate for both gram-negative and gram-positive bacteria.
Formel: C₂₁H₁₇N₃O₈S₃
MW: 535.57 g/mol
Storage Temperature: Gefrierschrank
CAS-Nummer: 80072-86-2
Order Now
Spezifikation Testergebnisse
Purity (HPLC) ≥98%
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge