Blocking buffer, fish gelatin
Lieferant: BIOTIUM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
22010 22011
22010.EA 79.2 EUR InStock
22010. 22011.
Blocking buffer, fish gelatin
Puffer Elektrophorese-Puffer Blockier-Puffer
Fish gelatin is commonly used in blocking solutions to minimise background noise in immunodetection procedures such as Western Blotting and immunostaining. Fish gelatin can be used to block non-specific binding sites on positively charged nylon or PVDF membranes or for blocking and dilution of antibodies during immunostaining procedures. Unlike BSA or milk, fish gelatin does not contain IgG or serum proteins that could cross-react with mammalian antibodies.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge