Tyramide amplification buffer plus
Lieferant: BIOTIUM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
22029
22029.EA 725 EUR InStock
22029.
Tyramide amplification buffer plus
Proteinkonjugate und Markierungen
An optimized buffer with enhanced sensitivity for tyramide signal amplification of immunostaining in cells or tissue sections.
  • For cell and tissue staining with tyramide signal amplification
  • Significantly improved brightness, specificity, and sensitivity
  • Compatible with CF® Dye and other tyramides
  • 250 or 1000 reaction sizes
  • 1× Tyramide amplification buffer plus
  • 30% Hydrogen peroxide
Tyramide amplification buffer plus is an optimized buffer with enhanced sensitivity for tyramide signal amplification of immunostaining in cells or tissue sections. It provides significantly improved brightness, specificity, and sensitivity compared to Biotium's original ready-to-use tyramide amplification buffer.
Tyramide amplification buffer plus was developed as a new and improved version of Biotium's original ready-to-use tyramide amplification buffer. It is also a component in all of Biotium's tyramide amplification kits.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge